1-Naphthylhydrazine hydrochloride ≥90.0% (by HPLC)

Supplier: TCI America

Synonyms: Naphthalen-1-ylhydrazine hydrochloride

N0064-1G N0064-5G
TCN0064-1GEA 33 USD
TCN0064-1G TCN0064-5G
1-Naphthylhydrazine hydrochloride ≥90.0% (by HPLC)
1-Naphthylhydrazine hydrochloride

CAS Number: 2243-56-3
MDL Number: MFCD00060157
Molecular Formula: C10H10N2
Molecular Weight: 194.66
Purity/Analysis Method: >90.0% (HPLC)
Form: Crystal

Formula: C₁₀H₁₀N₂
MW: 194.66 g/mol
MDL Number: MFCD00060157
CAS Number: 2243-56-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cf6633addf1e818a454ee5724e55a5ffd21885d8&vid=cf6633addf1e818a454ee5724e55a5ffd21885d8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >90.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR