Disodium-3-hydroxy-2,7-naphthalenedisulfonate ≥85.0% (by HPLC)
Supplier: TCI America
|
Warning
|
Synonyms:
2-Naphthol-3,6-disulphonic acid disodium salt, 3-Hydroxy-2,7-naphthalenedisulphonic acid disodium salt, Disodium 2-naphthol-3,6-disulphonate
N0029-25G
N0029-500G
TCN0029-025GEA
58
USD
TCN0029-025G
TCN0029-500G
Disodium-3-hydroxy-2,7-naphthalenedisulfonate ≥85.0% (by HPLC)
Disodium-3-hydroxy-2,7-naphthalenedisulfonate
CAS Number: 135-51-3
MDL Number: MFCD00065915
Molecular Formula: C10H8O7S2
Molecular Weight: 348.25
Purity/Analysis Method:
Other Product Names / Synonyms: Disodium 2-Naphthol-3,6-disulfonate, 3-Hydroxy-2,7-naphthalenedisulfonic Acid Disodium Salt, 2-Naphthol-3,6-disulfonic Acid Disodium Salt, R Acid Disodium Salt
Formula:
C₁₀H₆O₇S₂.2Na MW: 348.25 g/mol |
MDL Number:
MFCD00065915 CAS Number: 135-51-3 EINECS: 205-196-3 |
Specification Test Results
Appearance | White to Gray to Brown powder to crystal |
Purity (HPLC) | min. 85.0 area% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...