Sodium-1-naphthaleneacetate ≥96.0% (by HPLC, titration analysis)

Supplier: TCI America

N0007-25G N0007-500G
TCN0007-025GEA 24 USD
TCN0007-025G TCN0007-500G
Sodium-1-naphthaleneacetate ≥96.0% (by HPLC, titration analysis)
Sodium-1-naphthaleneacetate

CAS Number: 61-31-4
MDL Number: MFCD00064967
Molecular Formula: C12H10O2
Molecular Weight: 208.19
Purity/Analysis Method: >96.0% (HPLC,T)
Form: Crystal


Product Categories: Bioscience, Auxins
Other Product Names / Synonyms: 1-Naphthylacetic Acid Sodium Salt, 1-Naphthaleneacetic Acid Sodium Salt, Sodium 1-Naphthylacetate, Na-NAA

Formula: C₁₂H₁₀O₂
MDL Number: MFCD00064967
CAS Number: 61-31-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bff4453194fda8d80fbbd0784f4fffdce5438a8e&vid=bff4453194fda8d80fbbd0784f4fffdce5438a8e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR