6-Methyl-4-phenylchroman-2-one ≥98.0%

Supplier: TCI America

Synonyms: 6-Methyl-4-Phenylchromanone, 3,4-Dihydro-6-methyl-4-phenylcoumarin, 6-Methyl-4-phenyl-2-chromanone

M2093-25G M2093-5G
TCM2093-25GEA 116 USD
TCM2093-25G TCM2093-5G
6-Methyl-4-phenylchroman-2-one ≥98.0%
6-Methyl-4-phenylchroman-2-one

CAS Number: 40546-94-9
MDL Number: MFCD03695019
Molecular Formula: C16H14O2
Molecular Weight: 238.29
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 230
Melting point (°C): 82


Other Product Names / Synonyms: 3,4-Dihydro-6-methyl-4-phenylcoumarin

Formula: C₁₆H₁₄O₂
MW: 238.2812 g/mol
Boiling Pt: 230 °C
Melting Pt: 82 °C
MDL Number: MFCD03695019
CAS Number: 40546-94-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b6350005141708248c9978dd7a40cf88a93af1b&vid=4b6350005141708248c9978dd7a40cf88a93af1b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR