Methyl-3-bromo-2-(bromomethyl)propionate ≥97.0%

Supplier: TCI America

Synonyms: 3-Bromo-2-(bromomethyl)propionic Acid Methyl Ester, Methyl Bis(bromomethyl)acetate, Methyl beta,beta'-Dibromoisobutyrate, Methyl 3-bromo-2-(bromomethyl)propanoate

M2066-25G M2066-5G
TCM2066-25GEA 323 USD
TCM2066-25G TCM2066-5G
Methyl-3-bromo-2-(bromomethyl)propionate ≥97.0%
Methyl-3-bromo-2-(bromomethyl)propionate

CAS Number: 22262-60-8
MDL Number: MFCD00011534
Molecular Formula: C5H8Br2O2
Molecular Weight: 259.93
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 62
Flash Point (°C): 113
Specific Gravity (20/20): 1.85


Other Product Names / Synonyms: 3-Bromo-2-(bromomethyl)propionic Acid Methyl Ester, Methyl beta,beta'-Dibromoisobutyrate, Methyl Bis(bromomethyl)acetate

Formula: C₅H₈Br₂O₂
Boiling Pt: 62 °C
Density: 1.85 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00011534
CAS Number: 22262-60-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=849c04ace5d428411a6b5e1282b5362bb1bbfcde&vid=849c04ace5d428411a6b5e1282b5362bb1bbfcde&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR