1-Methyl-3-octylimidazolium hexafluorophosphate ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

M2063-25G M2063-5G
TCM2063-25GEA 242 USD
TCM2063-25G TCM2063-5G
1-Methyl-3-octylimidazolium hexafluorophosphate ≥98.0% (by HPLC, total nitrogen)
1-Methyl-3-octylimidazolium hexafluorophosphate

CAS Number: 304680-36-2
Molecular Formula: C12H23F6N2P
Molecular Weight: 340.29
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Clear Liquid
Specific Gravity (20/20): 1.24

Formula: C₁₂H₂₃F₆N₂P
MW: 340.29 g/mol
Density: 1.24 (20 °C)
MDL Number: MFCD06798203
CAS Number: 304680-36-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=252827a1141f7b496b3a0241a56c77880ce10f2b&vid=6328124c2f57236f30b8885b786d6200363eb370&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR