Mexiletine hydrochloride ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 1-(2,6-Dimethylphenoxy)propan-2-amine hydrochloride, 1-(2,6-Dimethylphenoxy)-2-propylamine Hydrochloride, Mexiletine HCl

M2040-1G M2040-5G
TCM2040-1GEA 39 USD
TCM2040-1G TCM2040-5G
Mexiletine hydrochloride ≥98.0% (by HPLC, total nitrogen)
Mexiletine hydrochloride

CAS Number: 5370-01-4
MDL Number: MFCD00216024
Molecular Formula: C11H17NO
Molecular Weight: 215.72
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 202
Lambda max.: 272 nm (MeOH)


Product Categories: Bioscience, Other Pharmaceutical Ingredients
Other Product Names / Synonyms: 1-(2,6-Dimethylphenoxy)-2-propylamine Hydrochloride

Formula: C₁₁H₁₇NO
MW: 215.72 g/mol
Melting Pt: 202 °C
MDL Number: MFCD00216024
CAS Number: 5370-01-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ad1ad0142af2270cddcb61dbaf7c0b828daae893&vid=ad1ad0142af2270cddcb61dbaf7c0b828daae893&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR