Methyl bromodifluoroacetate ≥97.0%

Supplier: TCI America

Synonyms: Bromodifluoroacetic Acid Methyl Ester

M2020-25G M2020-5G
TCM2020-25GEA 204 USD
TCM2020-25G TCM2020-5G
Methyl bromodifluoroacetate ≥97.0%
Methyl bromodifluoroacetate

CAS Number: 683-98-7
MDL Number: MFCD06248118
Molecular Formula: C3H3BrF2O2
Molecular Weight: 188.96
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 96
Specific Gravity (20/20): 1.72


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: Bromodifluoroacetic Acid Methyl Ester

Formula: C₃H₃BrF₂O₂
MW: 188.96 g/mol
Boiling Pt: 96 °C
Density: 1.72 (20 °C)
MDL Number: MFCD06248118
CAS Number: 683-98-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c3edf7d59dfbdad128706e3e295d90b781734a4e&vid=c3edf7d59dfbdad128706e3e295d90b781734a4e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR