7-Methoxy-4-quinolinol ≥97.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
4-Hydroxy-7-methoxyquinoline
M1966-1G
M1966-5G
TCM1966-1GEA
92
USD
TCM1966-1G
TCM1966-5G
7-Methoxy-4-quinolinol ≥97.0% (by titrimetric analysis)
7-Methoxy-4-quinolinol
CAS Number: 82121-05-9
MDL Number: MFCD00169015
Molecular Formula: C10H9NO2
Molecular Weight: 175.19
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 285
Other Product Names / Synonyms: 4-Hydroxy-7-methoxyquinoline
Formula:
C₁₀H₉NO₂ MW: 175.19 g/mol Melting Pt: 285 °C |
MDL Number:
MFCD00169015 CAS Number: 82121-05-9 |
Specification Test Results
Purity/Analysis Method | >97.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...