3-Maleimidopropionic acid ≥97.0%

Supplier: TCI America

Synonyms: 3-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)propanoic acid, 3-Maleimido-propionic acid, N-(2-Carboxyethyl)maleimide, MPS-Acid, N-Maleoyl-β-alanine

M1962-1G M1962-200MG
TCM1962-1GEA 90 USD
TCM1962-1G TCM1962-200MG
3-Maleimidopropionic acid ≥97.0%
3-Maleimidopropionic acid

CAS Number: 7423-55-4
MDL Number: MFCD00043030
Molecular Formula: C7H7NO4
Molecular Weight: 169.14
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 110

Formula: C₇H₇NO₄
MW: 169.14 g/mol
Melting Pt: 110 °C
MDL Number: MFCD00043030
CAS Number: 7423-55-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=94478430773bef3913b04f436db89dd9fb78900b&vid=94478430773bef3913b04f436db89dd9fb78900b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR