1-(4-Aminophenyl)-4-(4-methoxyphenyl)piperazine ≥96.0% (by HPLC, titration analysis)
Supplier: TCI America
M1920-25G
M1920-5G
TCM1920-25GEA
92
USD
TCM1920-25G
TCM1920-5G
1-(4-Aminophenyl)-4-(4-methoxyphenyl)piperazine ≥96.0% (by HPLC, titration analysis)
1-(4-Aminophenyl)-4-(4-methoxyphenyl)piperazine
CAS Number: 74852-62-3
MDL Number: MFCD02071496
Molecular Formula: C17H21N3O
Molecular Weight: 283.38
Purity/Analysis Method: >96.0% (HPLC,T)
Form: Crystal
Melting point (°C): 186
Other Product Names / Synonyms: 4-[4-(4-Methoxyphenyl)-1-piperazinyl]aniline
Formula:
C₁₇H₂₁N₃O Melting Pt: 186 °C |
MDL Number:
MFCD02071496 CAS Number: 74852-62-3 |
|
Specification Test Results
Purity/Analysis Method | >96.0% (HPLC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...