1-Methyl-L-glutamate ≥98.0%
Supplier: TCI America
Synonyms:
H-Glu-OMe, 1-Methyl L-Glutamate, L-Glutamic Acid 1-Methyl Ester
M1861-5G
M1861-1G
TCM1861-5GEA
157
USD
TCM1861-5G
TCM1861-1G
1-Methyl-L-glutamate ≥98.0%
1-Methyl-L-glutamate
CAS Number: 6384-08-3
MDL Number: MFCD00171677
Molecular Formula: C6H11NO4
Molecular Weight: 161.16
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 145
Specific rotation [a]20/D: 35 deg (C=2, H2O)
Product Categories: Bioscience, Amino Acids (C-Protected)
Other Product Names / Synonyms: L-Glutamic Acid 1-Methyl Ester, H-Glu-OMe
Formula:
C₆H₁₁NO₄ Melting Pt: 145 °C |
MDL Number:
MFCD00171677 CAS Number: 6384-08-3 |
|
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Specific rotation [a]20/D | +34.0 to +37.0 deg (C=2, H2O) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...