Methyl-3-(4-hydroxy-3-methoxyphenyl)propionate ≥98.0%
Supplier: TCI America
Synonyms:
3-(4-Hydroxy-3-methoxyphenyl)propionic Acid Methyl Ester, Methyl 3-(4-hydroxy-3-methoxyphenyl)propanoate
M1803-25G
M1803-5G
TCM1803-25GEA
743.8
USD
TCM1803-25G
TCM1803-5G
Methyl-3-(4-hydroxy-3-methoxyphenyl)propionate ≥98.0%
Methyl-3-(4-hydroxy-3-methoxyphenyl)propionate
CAS Number: 56024-44-3
Molecular Formula: C11H14O4
Molecular Weight: 210.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 146
Specific Gravity (20/20): 1.18
Other Product Names / Synonyms: 3-(4-Hydroxy-3-methoxyphenyl)propionic Acid Methyl Ester
Formula:
C₁₁H₁₄O₄ MW: 210.23 g/mol Boiling Pt: 146 °C Density: 1.18 (20 °C) |
MDL Number:
MFCD11975434 CAS Number: 56024-44-3 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...