3-Methoxybutyl-3-mercaptopropionate ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3-Mercaptopropionic Acid 3-Methoxybutyl Ester

M1802-25G M1802-500G
TCM1802-25GEA 17 USD
TCM1802-25G TCM1802-500G
3-Methoxybutyl-3-mercaptopropionate ≥97.0% (by GC, titration analysis)
3-Methoxybutyl-3-mercaptopropionate

CAS Number: 27431-40-9
MDL Number: MFCD04038137
Molecular Formula: C8H16O3S
Molecular Weight: 192.27
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Flash Point (°C): 126
Specific Gravity (20/20): 1.05


Product Categories: Materials Chemistry, Self-Assembled Monolayer (SAM) Forming Agents
Other Product Names / Synonyms: 3-Mercaptopropionic Acid 3-Methoxybutyl Ester

Formula: C₈H₁₆O₃S
MW: 192.27 g/mol
Density: 1.05 (20 °C)
Flash Pt: 126 °C
MDL Number: MFCD04038137
CAS Number: 27431-40-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6e47f7a00ba6903afcb6c34531b6ed0b11483b49&vid=6e47f7a00ba6903afcb6c34531b6ed0b11483b49&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR