2-(2-Methoxyphenoxy)ethanamine hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

M1794-5G M1794-25G
TCM1794-5GEA 25 USD
TCM1794-5G TCM1794-25G
2-(2-Methoxyphenoxy)ethanamine hydrochloride ≥98.0% (by titrimetric analysis)

CAS Number: 64464-07-9
MDL Number: MFCD01631120
Molecular Formula: C9H13NO2
Molecular Weight: 203.67
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Other Product Names / Synonyms: 2-(2-Aminoethoxy)anisole Hydrochloride, 1-(2-Aminoethoxy)-2-methoxybenzene Hydrochloride

Formula: C₉H₁₄ClNO₂
MDL Number: MFCD01631120
CAS Number: 64464-07-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e0f92e6d20d26e2db3670d8c6e6efccf5d587fc3&vid=e0f92e6d20d26e2db3670d8c6e6efccf5d587fc3&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR