4-Aminoantipyrine hydrochloride ≥98.0% (by HPLC, titration analysis) for biochemical research

Supplier: TCI America

Synonyms: 4-AA.HCl, ['4-AA·HCl'], 4-Aminoantipyrine Hydrochloride, 4-Amino-1,5-dimethyl-2-phenyl-1H-pyrazol-3(2H)-one hydrochloride

A0257-5G A0257-25G
TCA0257-5GEA 51.13 USD
TCA0257-5G TCA0257-025G
4-Aminoantipyrine hydrochloride ≥98.0% (by HPLC, titration analysis) for biochemical research
4-Aminoantipyrine hydrochloride

[for Biochemical Research]
CAS Number: 22198-72-7
MDL Number: MFCD00042012
Molecular Formula: C11H13N3O
Molecular Weight: 239.70
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal


Product Categories: Bioscience, Peroxidase Substrates
Other Product Names / Synonyms: 4-AA.HCl

Formula: C₁₁H₁₃N₃O
MW: 239.70 g/mol
MDL Number: MFCD00042012
CAS Number: 22198-72-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=771fd9b6198b330439c3ac75e61b8474e01bcc65&vid=0090556c7f89951d043fd4047e4cdda9c0d973ab&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR