Miconazole nitrate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

M1769-25G M1769-5G
TCM1769-25GEA 323 USD
TCM1769-25G TCM1769-5G
Miconazole nitrate ≥98.0% (by titrimetric analysis)
Miconazole nitrate

CAS Number: 22832-87-7
MDL Number: MFCD00058161
Molecular Formula: C18H14Cl4N2O
Molecular Weight: 479.14
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 184


Product Categories: Bioscience, Antifungal Ingredients
Other Product Names / Synonyms: 1-[2-(2,4-Dichlorobenzyloxy)-2-(2,4-dichlorophenyl)ethyl]-1H-imidazole Nitrate

Formula: C₁₈H₁₄Cl₄N₂O·HNO₃
Melting Pt: 184 °C
MDL Number: MFCD00058161
CAS Number: 22832-87-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b3e4b1fabf3b00bfc1622d43fd07e6d84cf16387&vid=b3e4b1fabf3b00bfc1622d43fd07e6d84cf16387&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR