N-Methyl-p-toluenesulfonamide ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: N,4-Dimethylbenzenesulphonamide

M1717-250G M1717-25G
TCM1717-250GEA 162.83 USD
TCM1717-250G TCM1717-25G
N-Methyl-p-toluenesulfonamide ≥98.0% (by HPLC, total nitrogen)
N-Methyl-p-toluenesulfonamide

CAS Number: 640-61-9
Molecular Formula: C8H11NO2S
Molecular Weight: 185.24
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 79

Formula: C₈H₁₁NO₂S
MW: 185.24 g/mol
Melting Pt: 79 °C
MDL Number: MFCD00008285
CAS Number: 640-61-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=47c2cafe38a54cc79f45643e09d31ba5610835fa&vid=47c2cafe38a54cc79f45643e09d31ba5610835fa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR