2'-Methyl-3'-nitroacetanilide ≥98.0%

Supplier: TCI America

Synonyms: N-(2-Methyl-3-nitrophenyl)acetamide

M1651-25G M1651-5G
TCM1651-25GEA 255.3 USD
TCM1651-25G TCM1651-5G
2'-Methyl-3'-nitroacetanilide ≥98.0%
2'-Methyl-3'-nitroacetanilide

CAS Number: 56207-36-4
Molecular Formula: C9H10N2O3
Molecular Weight: 194.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 162

Formula: C₉H₁₀N₂O₃
MW: 194.190 g/mol
Melting Pt: 162 °C
MDL Number: MFCD01047363
CAS Number: 56207-36-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ecb851ff262eb0d19924297ecc5e8aa6b26450c&vid=2ecb851ff262eb0d19924297ecc5e8aa6b26450c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR