4-Methoxyphenyl-3,4-O-isopropylidene-β-D-galactopyranoside ≥98.0%

Supplier: TCI America

M1593-1G M1593-5G
TCM1593-1GEA 115.1 USD
TCM1593-1G TCM1593-5G
4-Methoxyphenyl-3,4-O-isopropylidene-β-D-galactopyranoside ≥98.0%
4-Methoxyphenyl-3,4-O-isopropylidene-beta-D-galactopyranoside

CAS Number: 159922-67-5
MDL Number: MFCD06797135
Molecular Formula: C16H22O7
Molecular Weight: 326.35
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 151
Specific rotation [a]20/D: -18 deg (C=1, MeOH)
Storage Temperature: <0°C

Formula: C₁₆H₂₂O₇
Melting Pt: 151 °C
MDL Number: MFCD06797135
CAS Number: 159922-67-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=721855d10ea05adda79c36a7e662d1bca71ea2d1&vid=721855d10ea05adda79c36a7e662d1bca71ea2d1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR