2-Methylcyclopenta[l]phenanthrene ≥97.0%

Supplier: TCI America

M1411-100MG M1411-1G
TCM1411-100MGEA 123.4 USD
TCM1411-100MG TCM1411-1G
2-Methylcyclopenta[l]phenanthrene ≥97.0%
2-Methylcyclopenta[l]phenanthrene

CAS Number: 121254-39-5
MDL Number: MFCD03844801
Molecular Formula: C18H14
Molecular Weight: 230.31
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 166

Formula: C₁₈H₁₄
Melting Pt: 166 °C
MDL Number: MFCD03844801
CAS Number: 121254-39-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=007a168f0602cd813a442135bb52316de0b190ed&vid=007a168f0602cd813a442135bb52316de0b190ed&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR