3,3'-Methylenediindole ≥98.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: DIM, 3,3'-Diindolylmethane

M1399-25G M1399-5G
TCM1399-25GEA 171.6 USD
TCM1399-25G TCM1399-5G
3,3'-Methylenediindole ≥98.0% (by total nitrogen basis)
3,3'-Methylenediindole

CAS Number: 1968-05-4
MDL Number: MFCD00195766
Molecular Formula: C17H14N2
Molecular Weight: 246.31
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 167


Other Product Names / Synonyms: Di-3-indolylmethane

Formula: C₁₇H₁₄N₂
MW: 246.31 g/mol
Melting Pt: 167 °C
MDL Number: MFCD00195766
CAS Number: 1968-05-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9b8f5bee9a2aa4a668342ece96201e91364e0fdd&vid=9b8f5bee9a2aa4a668342ece96201e91364e0fdd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR