Benzyl-L-(+)-mandelate ≥98.0%

Supplier: TCI America

Synonyms: (S)-Benzyl 2-hydroxy-2-phenylacetate, L-(+)-Mandelic Acid Benzyl Ester

M1355-5G M1355-25G
TCM1355-005GEA 53 USD
TCM1355-005G TCM1355-025G
Benzyl-L-(+)-mandelate ≥98.0%
Benzyl-L-(+)-mandelate

CAS Number: 62173-99-3
MDL Number: MFCD00274325
Molecular Formula: C15H14O3
Molecular Weight: 242.27
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 107
Specific rotation [a]20/D: 34 deg (C=1, CH3CN)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: L-(+)-Mandelic Acid Benzyl Ester

Formula: C₁₅H₁₄O₃
MW: 242.27 g/mol
Melting Pt: 107 °C
MDL Number: MFCD00274325
CAS Number: 62173-99-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e69060cd6e808362ea21f074b2a8d3c5f095aac7&vid=e69060cd6e808362ea21f074b2a8d3c5f095aac7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR