(±)-2-Methylglutaric acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-Methylpentanedioic acid

M1300-25G M1300-5G
TCM1300-025GEA 235 USD
TCM1300-025G TCM1300-005G
(±)-2-Methylglutaric acid ≥98.0% (by GC, titration analysis)
(±)-2-Methylglutaric acid

CAS Number: 617-62-9
MDL Number: MFCD00002661
Molecular Formula: C6H10O4
Molecular Weight: 146.14
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 205
Melting point (°C): 77


Other Product Names / Synonyms: 2-Methylpentanedioic Acid

Formula: C₆H₁₀O₄
MW: 146.14 g/mol
Boiling Pt: 205 °C
Melting Pt: 77 °C
MDL Number: MFCD00002661
CAS Number: 617-62-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7155e9d6b318bbe1c1fa833c9e1bbb5d9c7f90f4&vid=7155e9d6b318bbe1c1fa833c9e1bbb5d9c7f90f4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR