4-Methoxybutyl acetate ≥98.0%

Supplier: TCI America

Synonyms: Acetic Acid 4-Methoxybutyl Ester

A1676-1G A1676-5G
TCA1676-1GEA 27 USD
TCA1676-1G TCA1676-5G
4-Methoxybutyl acetate ≥98.0%
4-Methoxybutyl acetate

CAS Number: 15057-11-1
MDL Number: MFCD06797048
Molecular Formula: C7H14O3
Molecular Weight: 146.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 71


Other Product Names / Synonyms: Acetic Acid 4-Methoxybutyl Ester

Formula: C₇H₁₄O₃
MW: 146.19 g/mol
Boiling Pt: 71 °C
MDL Number: MFCD06797048
CAS Number: 15057-11-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd0ec93a9b6170dcb11114e6e244bb37d5b2ecbf&vid=dd0ec93a9b6170dcb11114e6e244bb37d5b2ecbf&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR