2-Methyl-3-hexanol

Supplier: TCI America

M1266-5ML M1266-25ML
TCM1266-005MLEA 45 USD
TCM1266-005ML TCM1266-025ML
2-Methyl-3-hexanol
2-Methyl-3-hexanol

CAS Number: 617-29-8
MDL Number: MFCD00004577
Molecular Formula: C7H16O
Molecular Weight: 116.20
Form: Clear Liquid
Color: Colorless
Boiling point (°C): 145
Flash Point (°C): 41
Specific Gravity (20/20): 0.82

Formula: C₇H₁₆O
MW: 116.201 g/mol
Boiling Pt: 145 °C
Density: 0.82 (20 °C)
Flash Pt: 41 °C
MDL Number: MFCD00004577
CAS Number: 617-29-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d3fc30c35ae25c4200e5509e9ba1e45773e3fd38&vid=d3fc30c35ae25c4200e5509e9ba1e45773e3fd38&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR