2-Mesitylenesulfonyl hydrazide ≥97.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
2,4,6-Trimethylbenzenesulphonohydrazide, 2,4,6-Trimethylbenzenesulphonyl Hydrazide
M1198-25G
M1198-5G
TCM1198-025GEA
286.6
USD
TCM1198-025G
TCM1198-5G
2-Mesitylenesulfonyl hydrazide ≥97.0% (by titrimetric analysis)
2-Mesitylenesulfonyl hydrazide
CAS Number: 16182-15-3
MDL Number: MFCD00007585
Molecular Formula: C9H14N2O2S
Molecular Weight: 214.28
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 114
Other Product Names / Synonyms: 2,4,6-Trimethylbenzenesulfonyl Hydrazide
Formula:
C₉H₁₄N₂O₂S MW: 214.28 g/mol Melting Pt: 114 °C |
MDL Number:
MFCD00007585 CAS Number: 16182-15-3 |
Specification Test Results
Purity/Analysis Method | >97.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...