2,2,2-Trifluoroethyl methanesulfonate ≥98.0%
Supplier: TCI America
M1193-5G
M1193-25G
TCM1193-005GEA
17
USD
TCM1193-005G
TCM1193-25G
2,2,2-Trifluoroethyl methanesulfonate ≥98.0%
2,2,2-Trifluoroethyl methanesulfonate
CAS Number: 25236-64-0
MDL Number: MFCD00025058
Molecular Formula: C3H5F3O3S
Molecular Weight: 178.13
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 99
Flash Point (°C): 104
Specific Gravity (20/20): 1.52
Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Fluorinated Building Blocks, Other Fluorinated Group Introducing Agents
Other Product Names / Synonyms: Methanesulfonic Acid 2,2,2-Trifluoroethyl Ester
Formula:
C₃H₅F₃O₃S Boiling Pt: 99 °C Density: 1.52 (20 °C) Flash Pt: 104 °C |
MDL Number:
MFCD00025058 CAS Number: 25236-64-0 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...