3-Methylpyrrole ≥98.0%

Supplier: TCI America

Synonyms: 3-Methyl-1H-pyrrole

M1144-1G M1144-5G
TCM1144-001GEA 78.36 USD
TCM1144-001G TCM1144-5G
3-Methylpyrrole ≥98.0%
3-Methylpyrrole

CAS Number: 616-43-3
MDL Number: MFCD00083419
Molecular Formula: C5H7N
Molecular Weight: 81.12
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 144
Freezing point (°C): -48
Specific Gravity (20/20): 0.93

Formula: C₅H₇N
MW: 81.12 g/mol
Boiling Pt: 144 °C
Density: 0.93 (20 °C)
MDL Number: MFCD00083419
CAS Number: 616-43-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4548399e4d3f0e163af5be31953fe82bdb333e47&vid=4548399e4d3f0e163af5be31953fe82bdb333e47&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR