(3R)-(+)-3-(Methylamino)pyrrolidine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
(3R)-(+)-3-(Methylamino)pyrrolidine dihydrochloride
M1107-5G
M1107-1G
TCM1107-5GEA
675.8
USD
TCM1107-5G
TCM1107-001G
(3R)-(+)-3-(Methylamino)pyrrolidine ≥98.0% (by GC, titration analysis)
(3R)-(+)-3-(Methylamino)pyrrolidine
CAS Number: 139015-33-1
MDL Number: MFCD00191461
Molecular Formula: C5H12N2
Molecular Weight: 100.17
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 164
Specific Gravity (20/20): 0.94
Specific rotation [a]20/D: 19.5 deg (C=10, EtOH)
Storage Temperature: 0-10°C
Formula:
C₅H₁₂N₂ MW: 100.17 g/mol Boiling Pt: 164 °C Density: 0.94 (20 °C) |
MDL Number:
MFCD00191461 CAS Number: 139015-33-1 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...