Morpholinoacetonitrile ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: N-Cyanomethylmorpholine

M1095-25G M1095-5G
TCM1095-025GEA 356 USD
TCM1095-025G TCM1095-5G
Morpholinoacetonitrile ≥98.0% (by GC, titration analysis)
Morpholinoacetonitrile

CAS Number: 5807-02-3
MDL Number: MFCD00023372
Molecular Formula: C6H10N2O
Molecular Weight: 126.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 124
Melting point (°C): 62


Other Product Names / Synonyms: N-Cyanomethylmorpholine

Formula: C₆H₁₀N₂O
MW: 126.16 g/mol
Boiling Pt: 124 °C
Melting Pt: 62 °C
MDL Number: MFCD00023372
CAS Number: 5807-02-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4033cbecac4ecc0ed2ff03eadb993357af22355d&vid=4033cbecac4ecc0ed2ff03eadb993357af22355d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR