Methylsulfamic acid ≥98.0% (by titrimetric analysis)

Supplier: TCI America

M1083-25G M1083-5G
TCM1083-025GEA 783 USD
TCM1083-025G TCM1083-005G
Methylsulfamic acid ≥98.0% (by titrimetric analysis)
Methylsulfamic acid

CAS Number: 4112-03-2
MDL Number: MFCD00010870
Molecular Formula: CH5NO3S
Molecular Weight: 111.12
Purity/Analysis Method: >98.0% (T)
Form: Crystal

Formula: CH₅NO₃S
MW: 111.12 g/mol
MDL Number: MFCD00010870
CAS Number: 4112-03-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2a9df4f5ce270d1ddf8fb9aa32c6b7337bb82f14&vid=2a9df4f5ce270d1ddf8fb9aa32c6b7337bb82f14&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR