Monomyristin ≥97.0%
Supplier: TCI America
Synonyms:
1-Monomyristoyl Glycerol, Glycerol alpha-Monomyristate, 2,3-Dihydroxypropyl tetradecanoate
M1073-5G
M1073-1G
TCM1073-5GEA
99
USD
TCM1073-5G
TCM1073-001G
Monomyristin ≥97.0%
Monomyristin
CAS Number: 589-68-4
MDL Number: MFCD00046760
Molecular Formula: C17H34O4
Molecular Weight: 302.46
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 70
Product Categories: Materials Chemistry, Nonionic Surfactants, Bioscience, Lipids
Other Product Names / Synonyms: Glycerol alpha-Monomyristate, 1-Monomyristoyl Glycerol
Formula:
C₁₇H₃₄O₄ MW: 302.46 g/mol Melting Pt: 70 °C |
MDL Number:
MFCD00046760 CAS Number: 589-68-4 |
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...