Methyl Jasmonate (mixture of isomers) ≥90.0%

Supplier: TCI America

M1068-25G M1068-5G
TCM1068-25GEA 397 USD
TCM1068-25G TCM1068-005G
Methyl Jasmonate (mixture of isomers) ≥90.0%
Methyl Jasmonate (mixture of isomers)

CAS Number: 1101843-02-0
Molecular Formula: C13H20O3
Molecular Weight: 224.30
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 96
Flash Point (°C): 156
Specific Gravity (20/20): 1.02


Product Categories: Bioscience, Jasmonates
Other Product Names / Synonyms: Jasmonic Acid Methyl Ester, Methyl 3-Oxo-2-(2-pentenyl)cyclopentaneacetate, 3-Oxo-2-(2-pentenyl)cyclopentaneacetic Acid Methyl Ester, 2-(2-Pentenyl)-3-methoxycarbonylmethylcyclopentanone

Formula: C₁₃H₂₀O₃
Boiling Pt: 96 °C
Density: 1.02 (20 °C)
Flash Pt: 156 °C
CAS Number: 1101843-02-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=78753517345bfa7065561b2ab4e7df1c641dec8a&vid=78753517345bfa7065561b2ab4e7df1c641dec8a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >90.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR