Methyl-6-amino-2-naphthoate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 6-Amino-2-naphthoic acid methyll ester, Methyl 6-amino-2-naphthoate

A1643-1G A1643-5G
TCA1643-1GEA 151.7 USD
TCA1643-1G TCA1643-5G
Methyl-6-amino-2-naphthoate ≥98.0% (by GC, titration analysis)
Methyl 6-amino-2-naphthoate

CAS Number: 5159-59-1
MDL Number: MFCD06797040
Molecular Formula: C12H11NO2
Molecular Weight: 201.23
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 165


Other Product Names / Synonyms: 6-Amino-2-naphthoic Acid Methyl Ester

Formula: C₁₂H₁₁NO₂
MW: 201.2200 g/mol
Melting Pt: 165 °C
MDL Number: MFCD06797040
CAS Number: 5159-59-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c26797e90c72dfc8556e8d7a169b27b305526a53&vid=c26797e90c72dfc8556e8d7a169b27b305526a53&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR