Allyl ether ≥98.0%

Supplier: TCI America

Synonyms: Diallyl Ether

A0229-25ML A0229-500ML A0229-100ML
TCA0229-025MLEA 71.51 USD
TCA0229-025ML TCA0229-500ML TCA0229-100ML
Allyl ether ≥98.0%
Allyl ether

CAS Number: 557-40-4
MDL Number: MFCD00008650
Molecular Formula: C6H10O
Molecular Weight: 98.15
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 94
Flash Point (°C): -7
Specific Gravity (20/20): 0.80


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Diallyl Ether

Formula: C₆H₁₀O
Boiling Pt: 94 °C
Density: 0.80 (20 °C)
Flash Pt: –7 °C
MDL Number: MFCD00008650
CAS Number: 557-40-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=848c2f0c4cbbb418bd9b9e70898761cf49ef3e11&vid=848c2f0c4cbbb418bd9b9e70898761cf49ef3e11&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR