3-Methyl-1-cyclohexene ≥93.0%

Supplier: TCI America

M1034-500ML M1034-25ML
TCM1034-500MLEA 906 USD
TCM1034-500ML TCM1034-025ML
3-Methyl-1-cyclohexene ≥93.0%
3-Methyl-1-cyclohexene

CAS Number: 591-48-0
MDL Number: MFCD00001571
Molecular Formula: C7H12
Molecular Weight: 96.17
Purity/Analysis Method: >93.0% (GC)
Form: Clear Liquid
Boiling point (°C): 105
Flash Point (°C): -3
Specific Gravity (20/20): 0.80

Formula: C₇H₁₂
Boiling Pt: 105 °C
Melting Pt: –124 °C
Density: 0.80 (20 °C)
Flash Pt: –3 °C
MDL Number: MFCD00001571
CAS Number: 591-48-0
EINECS: 209-717-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d54a652add3c4f169c51afa9855710f6a7fb2a17&vid=d54a652add3c4f169c51afa9855710f6a7fb2a17&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >93.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR