2-Methyl-1-phenylpropene ≥98.0%

Supplier: TCI America

Synonyms: (2-Methylprop-1-en-1-yl)benzene, (2-Methylpropenyl)benzene

M1015-5G M1015-1G
TCM1015-005GEA 405.29 USD
TCM1015-005G TCM1015-001G
2-Methyl-1-phenylpropene ≥98.0%
2-Methyl-1-phenylpropene

CAS Number: 768-49-0
MDL Number: MFCD00008899
Molecular Formula: C10H12
Molecular Weight: 132.21
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 189
Flash Point (°C): 59
Specific Gravity (20/20): 0.90


Other Product Names / Synonyms: (2-Methylpropenyl)benzene

Formula: C₁₀H₁₂
MW: 132.21 g/mol
Boiling Pt: 189 °C
Density: 0.90 (20 °C)
Flash Pt: 59 °C
MDL Number: MFCD00008899
CAS Number: 768-49-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=51a9144d69a520e81742ba39ce8ce856081829d5&vid=51a9144d69a520e81742ba39ce8ce856081829d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR