4-Methoxy-2-nitrobenzenesulfonyl chloride ≥93.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 4-Methoxy-2-nitrobenzene-1-sulphonyl chloride

M1006-25G M1006-5G
TCM1006-025GEA 148.6 USD
TCM1006-025G TCM1006-5G
4-Methoxy-2-nitrobenzenesulfonyl chloride ≥93.0% (by titrimetric analysis)
4-Methoxy-2-nitrobenzenesulphonyl chloride

CAS Number: 18092-54-1
MDL Number: MFCD00143333
Molecular Formula: C7H6ClNO5S
Molecular Weight: 251.64
Purity/Analysis Method: >93.0% (T)
Form: Crystal
Melting point (°C): 77

Formula: C₇H₆ClNO₅S
MW: 251.65 g/mol
Melting Pt: 77 °C
MDL Number: MFCD00143333
CAS Number: 18092-54-1
EINECS: 241-996-9
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=906855d8f8fb9bd93cf8b504d82e2b9e12ab83d5&vid=906855d8f8fb9bd93cf8b504d82e2b9e12ab83d5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >93.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR