Ethyl-3-methoxypropionate ≥99.0%
Supplier: TCI America
Synonyms:
3-Methoxypropionic acid ethyl ester
M1001-10G
M1001-25G
TCM1001-010GEA
45
USD
TCM1001-010G
TCM1001-025G
Ethyl-3-methoxypropionate ≥99.0%
Ethyl 3-methoxypropionate
CAS Number: 10606-42-5
MDL Number: MFCD00143172
Molecular Formula: C6H12O3
Molecular Weight: 132.16
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 158
Specific Gravity (20/20): 0.98
Other Product Names / Synonyms: 3-Methoxypropionic Acid Ethyl Ester
Formula:
C₆H₁₂O₃ Boiling Pt: 158 °C Density: 0.98 (20 °C) |
MDL Number:
MFCD00143172 CAS Number: 10606-42-5 |
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...