5-Methyl-2-heptene (cis- and trans- mixture) ≥95.0%

Supplier: TCI America

M0992-1ML M0992-5ML
TCM0992-001MLEA 376.6 USD
TCM0992-001ML TCM0992-5ML
5-Methyl-2-heptene (cis- and trans- mixture) ≥95.0%
5-Methyl-2-heptene (cis- and trans- mixture)

CAS Number: 22487-87-2
MDL Number: MFCD00061016
Molecular Formula: C8H16
Molecular Weight: 112.22
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Flash Point (°C): 13
Specific Gravity (20/20): 0.73

Formula: C₈H₁₆
Density: 0.73 (20 °C)
Flash Pt: 13 °C
MDL Number: MFCD00061016
CAS Number: 22487-87-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=775a1802a5ebd10a1c8956c544be9c78751b0330&vid=775a1802a5ebd10a1c8956c544be9c78751b0330&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR