Benzyl methyl malonate ≥96.0%

Supplier: TCI America

M0974-25G M0974-5G
TCM0974-025GEA 161 USD
TCM0974-025G TCM0974-5G
Benzyl methyl malonate ≥96.0%
Benzyl methyl malonate

CAS Number: 52267-39-7
MDL Number: MFCD00008461
Molecular Formula: C11H12O4
Molecular Weight: 208.21
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Flash Point (°C): 110
Specific Gravity (20/20): 1.16


Other Product Names / Synonyms: Malonic Acid Benzyl Methyl Ester

Formula: C₁₁H₁₂O₄
Density: 1.16 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00008461
CAS Number: 52267-39-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7a5eaab586b022cff11a9991ee9468d37fd7f180&vid=7a5eaab586b022cff11a9991ee9468d37fd7f180&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR