2-(2-Hydroxyethyl)-1-methylpyrrolidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 1-Methyl-2-pyrrolidineethanol

M0908-10ML M0908-25ML
TCM0908-010MLEA 90 USD
TCM0908-010ML TCM0908-025ML
2-(2-Hydroxyethyl)-1-methylpyrrolidine ≥98.0% (by GC, titration analysis)
2-(2-Hydroxyethyl)-1-methylpyrrolidine

CAS Number: 67004-64-2
MDL Number: MFCD00003174
Molecular Formula: C7H15NO
Molecular Weight: 129.20
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 111
Flash Point (°C): 84
Specific Gravity (20/20): 0.96


Other Product Names / Synonyms: 1-Methyl-2-pyrrolidineethanol

Formula: C₇H₁₅NO
MW: 129.20 g/mol
Boiling Pt: 111 °C
Density: 0.96 (20 °C)
Flash Pt: 84 °C
MDL Number: MFCD00003174
CAS Number: 67004-64-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=218707dce176d866a354a5122245f4a6e5116c80&vid=218707dce176d866a354a5122245f4a6e5116c80&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR