Methyl propargyl ether ≥98.0%
Supplier: TCI America
Synonyms:
Methyl 2-Propynyl Ether
M0860-25ML
M0860-5ML
TCM0860-025MLEA
276
USD
TCM0860-025ML
TCM0860-5ML
Methyl propargyl ether ≥98.0%
Methyl propargyl ether
CAS Number: 627-41-8
MDL Number: MFCD00008578
Molecular Formula: C4H6O
Molecular Weight: 70.09
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 62
Flash Point (°C): -18
Specific Gravity (20/20): 0.85
Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry), Building Blocks ([2+2+2] Cycloaddition Reaction)
Other Product Names / Synonyms: Methyl 2-Propynyl Ether
Formula:
C₄H₆O MW: 70.09 g/mol Boiling Pt: 62 °C Density: 0.85 (20 °C) Flash Pt: –18 °C |
MDL Number:
MFCD00008578 CAS Number: 627-41-8 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...