2-Methoxyphenethylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

M0844-25ML M0844-5ML
TCM0844-025MLEA 145 USD
TCM0844-025ML TCM0844-5ML
2-Methoxyphenethylamine ≥98.0% (by GC, titration analysis)
2-Methoxyphenethylamine

CAS Number: 2045-79-6
MDL Number: MFCD00008186
Molecular Formula: C9H13NO
Molecular Weight: 151.21
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 237
Flash Point (°C): 113
Specific Gravity (20/20): 1.04


Product Categories: Bioscience, Phenylethylamines
Other Product Names / Synonyms: 2-(2-Aminoethyl)anisole, 2-Methoxyphenethylamine

Formula: C₉H₁₃NO
MW: 151.21 g/mol
Boiling Pt: 237 °C
Density: 1.04 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00008186
CAS Number: 2045-79-6
EINECS: 218-066-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=31899be9bebf37ee56e7066a654ae5c4e5ab8699&vid=31899be9bebf37ee56e7066a654ae5c4e5ab8699&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR