Allylamine hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Allylamine Hydrochloride

A0226-500G A0226-25G A0226-100G
TCA0226-500GEA 664.17 USD
TCA0226-500G TCA0226-025G TCA0226-100G
Allylamine hydrochloride ≥98.0% (by titrimetric analysis)
Allylamine hydrochloride

CAS Number: 10017-11-5
MDL Number: MFCD00054329
Molecular Formula: C3H7N
Molecular Weight: 93.55
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 106


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 3-Aminopropene Hydrochloride

Formula: C₃H₈ClN
MW: 93.5553 g/mol
Melting Pt: 106 °C
MDL Number: MFCD00054329
CAS Number: 10017-11-5
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=83c5a843151aed3b256a22212e9ee01403dbbb66&vid=83c5a843151aed3b256a22212e9ee01403dbbb66&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR