4-Methoxyphenethylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

M0795-25ML M0795-250ML
TCM0795-025MLEA 89 USD
TCM0795-025ML TCM0795-250ML
4-Methoxyphenethylamine ≥98.0% (by GC, titration analysis)
4-Methoxyphenethylamine

CAS Number: 55-81-2
MDL Number: MFCD00008192
Molecular Formula: C9H13NO
Molecular Weight: 151.21
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 138
Flash Point (°C): 110
Specific Gravity (20/20): 1.03


Other Product Names / Synonyms: 4-(2-Aminoethyl)anisole, 4-Methoxyphenethylamine

Formula: C₉H₁₃NO
MW: 151.21 g/mol
Boiling Pt: 138 °C
Density: 1.03 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00008192
CAS Number: 55-81-2
EINECS: 200-245-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=96f68c22acdfc9348f2a43bcdebb04a424c4795f&vid=96f68c22acdfc9348f2a43bcdebb04a424c4795f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR