(4-Methoxyphenyl)acetic acid ethyl ester ≥98.0%

Supplier: TCI America

Synonyms: Ethyl (4-methoxyphenyl)acetate, Ethyl 2-(4-methoxyphenyl)acetate, Homoanisic acid ethyl ester, 4-Methoxyphenylacetic acid ethyl ester, Ethyl 4-Methoxyphenylacetate

M0793-25ML M0793-500ML
TCM0793-025MLEA 88 USD
TCM0793-025ML TCM0793-500ML
(4-Methoxyphenyl)acetic acid ethyl ester ≥98.0%
(4-Methoxyphenyl)acetic acid ethyl ester

CAS Number: 14062-18-1
MDL Number: MFCD00040760
Molecular Formula: C11H14O3
Molecular Weight: 194.23
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 158
Specific Gravity (20/20): 1.08


Other Product Names / Synonyms: 4-Methoxyphenylacetic Acid Ethyl Ester, Homoanisic Acid Ethyl Ester

Formula: C₁₁H₁₄O₃
MW: 194.23 g/mol
Boiling Pt: 158 °C
Density: 1.08 (20 °C)
MDL Number: MFCD00040760
CAS Number: 14062-18-1
EINECS: 237-903-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73f4491e7154bee648df0718d657ed684b111f51&vid=73f4491e7154bee648df0718d657ed684b111f51&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR