3-Methyl-2-oxazolidone ≥99.0%
Supplier: TCI America
M0781-25G
M0781-500G
M0781-100G
TCM0781-025GEA
72.47
USD
TCM0781-025G
TCM0781-500G
TCM0781-100G
3-Methyl-2-oxazolidone ≥99.0%
3-Methyl-2-oxazolidone
CAS Number: 19836-78-3
MDL Number: MFCD00009759
Molecular Formula: C4H7NO2
Molecular Weight: 101.11
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 88
Flash Point (°C): 110
Specific Gravity (20/20): 1.18
Other Product Names / Synonyms: 2-Oxo-3-methyloxazolidine
Formula:
C₄H₇NO₂ Boiling Pt: 88 °C Density: 1.18 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00009759 CAS Number: 19836-78-3 |
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...