1-Methoxy-3-(trimethylsilyloxy)-1,3-butadiene ≥95.0%

Supplier: TCI America

Synonyms: Danishefsky's Diene

M0759-5ML M0759-25ML
TCM0759-005MLEA 273 USD
TCM0759-005ML TCM0759-25ML
1-Methoxy-3-(trimethylsilyloxy)-1,3-butadiene ≥95.0%
1-Methoxy-3-(trimethylsilyloxy)-1,3-butadiene

CAS Number: 59414-23-2
MDL Number: MFCD00008498
Molecular Formula: C8H16O2Si
Molecular Weight: 172.30
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Flash Point (°C): 45
Specific Gravity (20/20): 0.90
Storage Temperature: 0-10°C


Product Categories: Synthetic Organic Chemistry, Silicon Compounds (for Synthesis)
Other Product Names / Synonyms: Danishefsky's Diene

Formula: C₈H₁₆O₂Si
Density: 0.90 (20 °C)
Flash Pt: 45 °C
MDL Number: MFCD00008498
CAS Number: 59414-23-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9579039e961b722eaf19b1c980c726c576b9f3b0&vid=9579039e961b722eaf19b1c980c726c576b9f3b0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR